Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153547.6 | NP_705775.2 | 12 | 1455 | ||
GenBank mRNA | AA041888.1 | - | 26 | |||
AK077523.1 | - | 12 | 1386 | |||
AK167183.1 | - | 12 | 1339 | |||
AY181025.1 | - | 12 | 1313 | |||
AY185498.1 | - | 12 | 1313 | |||
BC018560.1 | - | 12 | 1294 | |||
BC037996.1 | - | 12 | 1315 | 5052858 |