Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015767.3 | NP_056582.1 | 4, 5 | 682 | ||
GenBank mRNA | AF218416.1 | - | 4, 5 | 670 | ||
AK004882.1 | - | 4, 5 | 663 | |||
AK050374.1 | - | 4, 5 | 683 | |||
BC026882.1 | - | 4, 5 | 569 | |||
BC099868.1 | - | 4, 5 | 681 | 40040166 | ||
BC099869.1 | - | 4, 5 | 683 | 40040167 | ||
BC099870.1 | - | 4, 5 | 683 | 40040169 | ||
BC099871.1 | - | 4, 5 | 682 |