Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap30l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081168.1 | NP_001074637.1 | 4 | 903 | ||
GenBank mRNA | AK010234.1 | - | 4 | 391 | ||
AK203234.1 | - | 2 | 91 | |||
BC051686.1 | - | 4 | 903 | 6433799 | ||
BC094930.1 | - | 5 | 622 | |||
BC117029.1 | - | 4 | 617 | 40126348 | ||
BC117031.1 | - | 4 | 617 | 40126350 |