Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293667.1 | NP_001280596.1 | 20, 21 | 2733 | ||
NM_001293668.1 | NP_001280597.1 | 20, 21 | 2733 | |||
NM_015804.3 | NP_056619.1 | 20, 21 | 2733 | |||
GenBank mRNA | AF156551.1 | - | 20, 21 | 2525 | ||
AK147497.1 | - | 20, 21 | 2736 | |||
AK173088.1 | - | 20, 21 | 2886 | |||
BC036322.1 | - | 5, 6 | 794 | |||
BC138715.1 | - | 20, 21 | 2452 | 8861737 | ||
BC145399.1 | - | 20, 21 | 2453 | |||
BC145400.1 | - | 20, 21 | 2453 | |||
CF167530.1 | - | 2, 3 | 273 |