Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015825.2 | NP_056640.1 | 3 | 485 | ||
GenBank mRNA | AJ239082.1 | - | 3 | 251 | ||
AJ272170.1 | - | 3 | 313 | |||
AJ487014.1 | - | 3 | 312 | |||
BC055870.1 | - | 3 | 302 |