Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016703.3 | NP_057912.2 | 8 | 1258 | ||
GenBank mRNA | AF150808.1 | - | 8 | 1161 | ||
AF193017.1 | - | 8 | 1170 | |||
AK029838.1 | - | 8 | 1194 | |||
AK036866.1 | - | 8 | 1196 | |||
AK151288.1 | - | 8 | 1240 | |||
AK202976.1 | - | 3 | 153 | |||
AK206719.1 | - | 3 | 214 | |||
BC017527.1 | - | 8 | 1161 | 4921119 |