Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145528.3 | NP_663503.1 | 12 | 1354 | ||
GenBank mRNA | AK122339.1 | - | 8 | 3716 | ||
AK149672.1 | - | 12 | 1313 | |||
AK154470.1 | - | 12 | 1342 | |||
AK163962.1 | - | 12 | 1308 | |||
AK171206.1 | - | 12 | 1340 | |||
AK171440.1 | - | 12 | 1331 | |||
AK181445.1 | - | 2 | 90 | |||
AK184073.1 | - | 1 | 91 | |||
AK184342.1 | - | 2 | 186 | |||
BC016669.1 | - | 12 | 1338 | 2648797 | ||
BC023673.1 | - | 12 | 1304 | 5343236 | ||
BC023712.1 | - | 12 | 1327 | 5346303 | ||
BC033419.1 | - | 12 | 1305 | 5359944 |