Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162538.1 | NP_001156010.1 | 12 | 1451 | ||
NM_001162539.1 | NP_001156011.1 | 11 | 1322 | |||
NM_025714.4 | NP_079990.2 | 12 | 1317 | |||
GenBank mRNA | AK014630.1 | - | 12 | 1319 | ||
AK016726.1 | - | 12 | 1433 | |||
AK031564.1 | - | 12 | 1453 | |||
AK220159.1 | - | 2 | 140 | |||
BC020075.1 | - | 11 | 1291 | 3979963 |