Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033560.3 | NP_291038.2 | 6 | 995 | ||
GenBank mRNA | AK016385.1 | - | 6 | 978 | ||
AK048942.1 | - | 6 | 975 | |||
AK054055.1 | - | 6 | 977 | |||
AK137479.1 | - | 6 | 980 | |||
AK157507.1 | - | 6 | 962 | |||
AK170280.1 | - | 6 | 964 | |||
AK170943.1 | - | 6 | 953 | |||
AK170961.1 | - | 6 | 984 | |||
AK171055.1 | - | 6 | 984 | |||
AK171221.1 | - | 6 | 996 | |||
BC039290.1 | - | 6 | 951 | 5066508 | ||
BC110396.1 | - | 6 | 910 | 5253683 |