Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026740.2 | NP_081016.2 | 2 | 906 | ||
GenBank mRNA | AK003278.1 | - | 2 | 889 | ||
AK170505.1 | - | 2 | 899 | |||
AK220201.1 | - | 2 | 866 | |||
BC024522.1 | - | 1 | 42 | |||
BC057976.1 | - | 2 | 891 | 5052851 |