Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002261.2 | NP_002252.2 | 1 | 110 | ||
NM_007333.2 | NP_031359.2 | 1 | 110 |