Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023175.1 | NP_075664.1 | 5 | 418 | ||
GenBank mRNA | AF284573.1 | - | 5 | 418 | ||
AK003604.1 | - | 2 | 113 | |||
AK004533.1 | - | 5 | 402 | |||
AK004535.1 | - | 5 | 404 | |||
BC020153.1 | - | 5 | 407 | 3992930 |