Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290697.1 | NP_001277626.1 | 3 | 606 | ||
NM_026023.5 | NP_080299.4 | 3 | 681 | |||
GenBank mRNA | AK004388.1 | - | 3 | 675 | ||
AK005909.1 | - | 3 | 676 | |||
AK010850.1 | - | 3 | 488 | |||
AK012388.1 | - | 3 | 459 | |||
AK031779.1 | - | 3 | 459 | |||
BC005646.1 | - | 3 | 442 | 3708975 | ||
BE949482.1 | - | 3 | 326 |