Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114396.1 | NP_001107868.1 | 4 | 345 | ||
NM_002262.3 | NP_002253.1 | 3 | 408 | |||
NM_007334.2 | NP_031360.1 | 2 | 315 | |||
XM_006719067.3 | XP_006719130.1 | 3 | 397 | |||
XM_011520650.2 | XP_011518952.1 | 4 | 339 | |||
XM_011520651.2 | XP_011518953.1 | 3 | 396 | |||
XM_017019285.1 | XP_016874774.1 | 3 | 400 | |||
XM_017019286.1 | XP_016874775.1 | 4 | 330 | |||
XM_017019287.1 | XP_016874776.1 | 4 | 350 | |||
XM_017019288.1 | XP_016874777.1 | 2 | 318 | |||
XM_017019289.1 | XP_016874778.1 | 2 | 312 | |||
XM_017019290.1 | XP_016874779.1 | 5 | 2206 | |||
XM_017019291.1 | XP_016874780.1 | 4 | 332 | |||
GenBank mRNA | AB009597.1 | - | 2 | 92 | ||
AF498040.1 | - | 3 | 276 | |||
AF498041.1 | - | 3 | 276 | |||
AJ000001.1 | - | 3 | 176 | |||
AK291506.1 | - | 4 | 350 | |||
AY227806.1 | - | 4 | 345 | |||
BC028009.1 | - | 3 | 221 | 5213081 | ||
U30610.1 | - | 3 | 232 |