Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252582.1 | NP_001239511.1 | 8 | 1486 | ||
NM_001252583.1 | NP_001239512.1 | 8 | 1451 | |||
NM_144833.3 | NP_659082.1 | 8 | 1451 | |||
GenBank mRNA | AF295806.1 | - | 8 | 1213 | ||
AK033090.1 | - | 8 | 1228 | |||
AK042127.1 | - | 1 | 684 | |||
AK051939.1 | - | 6 | 851 | |||
AK053001.1 | - | 8 | 1176 | |||
AK083003.1 | - | 8 | 1194 | |||
AK132345.1 | - | 8 | 1189 | |||
BC021528.1 | - | 8 | 1206 | 5353673 | ||
BC052080.1 | - | 8 | 1183 | 6403411 | ||
BC056957.1 | - | 8 | 1148 |