Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnlz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139504.1 | NP_001132976.1 | 2 | 443 | ||
NM_026828.3 | NP_081104.1 | 2 | 439 | |||
GenBank mRNA | AK004089.1 | - | 2 | 429 | ||
AK142070.1 | - | 2 | 440 | |||
BC139333.1 | - | 2 | 403 | 8862355 | ||
BC139334.1 | - | 2 | 402 | 8862356 |