Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rgs7bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029879.2 | NP_084155.2 | 3, 4 | 1172 | ||
GenBank mRNA | AK020910.1 | - | 3, 4 | 769 | ||
AK028323.1 | - | 3, 4 | 549 | |||
AK032138.1 | - | 3, 4 | 1172 | |||
AK036240.1 | - | 3, 4 | 1079 | |||
AK046733.1 | - | 3, 4 | 801 | |||
AK137417.1 | - | 3, 4 | 1046 | |||
BC118957.2 | - | 3, 4 | 704 | 40093423 | ||
DQ104214.1 | - | 3, 4 | 462 |