Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002263.3 | NP_002254.2 | 10 | 2408 | ||
XM_011514585.1 | XP_011512887.1 | 10 | 2114 | |||
XM_011514587.2 | XP_011512889.1 | 9 | 1334 | |||
XM_017010836.1 | XP_016866325.1 | 10 | 2114 | |||
XM_017010837.1 | XP_016866326.1 | 10 | 2223 | |||
GenBank mRNA | AK297378.1 | - | 10 | 2021 | ||
AK303239.1 | - | 10 | 2048 | |||
BC000712.2 | - | 10 | 2116 | |||
BC063567.1 | - | 10 | 2066 | |||
BC073878.1 | - | 10 | 2101 | |||
BC098438.1 | - | 10 | 2098 | |||
BC121041.1 | - | 10 | 2039 | 40117894 | ||
BC121042.1 | - | 10 | 2039 | 40117899 | ||
D14678.1 | - | 5 | 1498 |