Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026203.3 | NP_080479.2 | 10 | 1915 | ||
GenBank mRNA | AK036336.1 | - | 10 | 1805 | ||
AY133241.1 | - | 10 | 1808 | |||
BC038502.1 | - | 3 | 266 | |||
BC055400.1 | - | 9 | 1637 | 6467369 | ||
BC065146.1 | - | 10 | 1811 |