Search Thermo Fisher Scientific
Gene Symbol: KIF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005355.3 | NP_005346.3 | 6 | 871 | ||
NM_030615.2 | NP_085118.2 | 6 | 871 | |||
XM_011535802.2 | XP_011534104.1 | 8 | 978 | |||
XM_011535803.2 | XP_011534105.1 | 8 | 978 | |||
GenBank mRNA | AB012722.2 | - | 6 | 871 | ||
AK289488.1 | - | 8 | 1124 | |||
BC130489.1 | - | 6 | 919 |