Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281929.1 | NP_001268858.1 | 11 | 2053 | ||
NM_001290451.1 | NP_001277380.1 | 11 | 1825 | |||
NM_016886.4 | NP_058582.3 | 11 | 2053 | |||
GenBank mRNA | AB022342.1 | - | 11 | 1843 | ||
AB079072.1 | - | 11 | 1789 | |||
AB102775.1 | - | 11 | 1789 | |||
AB102776.1 | - | 11 | 1789 | |||
AF483494.1 | - | 11 | 1789 | |||
AF483495.1 | - | 11 | 1789 | |||
AK054154.1 | - | 1 | 1458 | |||
AK135043.1 | - | 11 | 1807 | |||
AK138615.1 | - | 11 | 1958 | |||
AK157815.1 | - | 12 | 2230 | |||
AK161163.1 | - | 11 | 1873 | |||
AK220530.1 | - | 11 | 1840 | |||
BC076584.1 | - | 11 | 1917 | |||
BC086678.1 | - | 11 | 1998 | 30668476 | ||
BC129855.1 | - | 11 | 1843 | 40109751 | ||
BC129856.1 | - | 11 | 1843 | 40109752 |