Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002268.4 | NP_002259.1 | 12 | 1244 | ||
GenBank mRNA | AB002533.2 | - | 12 | 947 | ||
AK291041.1 | - | 12 | 1196 | |||
BC028691.2 | - | 12 | 1189 | 4838709 | ||
BC034493.2 | - | 12 | 1244 | 4821602 | ||
U93240.1 | - | 12 | 1088 | |||
Y12393.1 | - | 3 | 218 |