Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002269.2 | NP_002260.2 | 5 | 492 | ||
XM_011535805.1 | XP_011534107.1 | 6 | 670 | |||
XM_017010838.1 | XP_016866327.1 | 6 | 670 | |||
XM_017010839.1 | XP_016866328.1 | 5 | 473 | |||
XM_017010840.1 | XP_016866329.1 | 6 | 630 | |||
XM_017010842.1 | XP_016866331.1 | 5 | 473 | |||
XM_017010843.1 | XP_016866332.1 | 6 | 597 | |||
GenBank mRNA | AF005361.1 | - | 4 | 357 | ||
AK314206.1 | - | 5 | 485 | |||
BC047409.1 | - | 5 | 484 | 5297277 | ||
BG721781.1 | - | 6 | 618 |