Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016919.2 | NP_058615.1 | 12, 13 | 1408 | ||
GenBank mRNA | AB040491.1 | - | 12, 13 | 1412 | ||
AF176645.1 | - | 12, 13 | 1408 | |||
BC138417.1 | - | 12, 13 | 1373 | 8861437 | ||
BC138419.1 | - | 12, 13 | 1373 | 8861439 | ||
BC145269.1 | - | 12, 13 | 1369 |