Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017394.4 | NP_059090.3 | 3 | 539 | ||
GenBank mRNA | AB026688.1 | - | 3 | 453 | ||
AK031867.1 | - | 3 | 507 | |||
BC054765.1 | - | 3 | 482 | 6414759 |