Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289653.1 | NP_001276582.1 | 4 | 281 | ||
GenBank mRNA | AK035019.1 | - | 2 | 134 | ||
AK039790.1 | - | 345 | ||||
AK048913.1 | - | 552 | ||||
BC051968.1 | - | 3 | 249 | 6401465 | ||
BU612839.1 | - | 4 | 490 |