Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017471.2 | NP_059499.1 | 4 | 337 | ||
GenBank mRNA | AF005204.1 | - | 4 | 341 | ||
AK137501.1 | - | 4 | 364 |