Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161796.1 | NP_001155268.1 | 10 | 1521 | ||
NM_017469.4 | NP_059497.1 | 10 | 1521 | |||
GenBank mRNA | AF020339.1 | - | 10 | 1495 | ||
AF297083.1 | - | 10 | 1495 | |||
AK139406.1 | - | 10 | 1484 | |||
AK147128.1 | - | 10 | 1456 | |||
BC050945.1 | - | 10 | 1499 | 6822142 | ||
BC056995.1 | - | 10 | 1499 | 6822142 |