Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017472.4 | NP_059500.2 | 3 | 623 | ||
GenBank mRNA | AF062482.1 | - | 3 | 421 | ||
AK007798.1 | - | 3 | 434 | |||
AK013183.1 | - | 3 | 648 | |||
AK013261.1 | - | 3 | 256 | |||
AK018790.1 | - | 3 | 256 | |||
AK145350.1 | - | 3 | 426 | |||
AK154728.1 | - | 3 | 426 | |||
AK188907.1 | - | 52 | ||||
BC116885.1 | - | 3 | 467 | 40126204 | ||
BC116889.1 | - | 3 | 467 | 40126208 |