Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000223.3 | NP_000214.1 | 1 | 551 | ||
GenBank mRNA | AK313747.1 | - | 1 | 565 | ||
D78367.1 | - | 1 | 551 |