Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138603.3 | NP_613069.3 | 12 | 1541 | ||
GenBank mRNA | AF229637.1 | - | 12 | 1482 | ||
AK010536.1 | - | 12 | 1459 | |||
AK039573.1 | - | 12 | 1514 | |||
BC011195.1 | - | 12 | 1446 | 4188459 | ||
BC057686.1 | - | 3 | 1591 |