Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018864.5 | NP_061352.2 | 8 | 751 | ||
GenBank mRNA | AF042730.1 | - | 8 | 696 | ||
AK011770.1 | - | 8 | 689 | |||
AK078406.1 | - | 8 | 700 | |||
AK083238.1 | - | 8 | 685 | |||
AK145472.1 | - | 688 | ||||
AK147778.1 | - | 8 | 711 | |||
AK152774.1 | - | 8 | 702 | |||
AK153457.1 | - | 8 | 718 | |||
AK160203.1 | - | 8 | 689 | |||
AK160467.1 | - | 8 | 683 | |||
AK165977.1 | - | 8 | 693 | |||
AK168457.1 | - | 8 | 726 | |||
BC049080.1 | - | 8 | 692 | 6419835 | ||
BC055722.1 | - | 7 | 646 | 6413389 |