Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pigb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018889.3 | NP_061377.2 | 10 | 1368 | ||
GenBank mRNA | AK153819.1 | - | 10 | 1285 | ||
BC052658.1 | - | 10 | 1328 | 30048340 | ||
D84436.1 | - | 10 | 1368 |