Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018859.2 | NP_061347.2 | 5 | 552 | ||
GenBank mRNA | AK002507.1 | - | 5 | 527 | ||
AK007924.1 | - | 5 | 529 | |||
AK028605.1 | - | 378 | ||||
U68535.1 | - | 5 | 538 |