Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019518.3 | NP_062391.3 | 2, 3 | 396 | ||
GenBank mRNA | AB041603.1 | - | 2, 3 | 394 | ||
AF236099.1 | - | 2, 3 | 359 | |||
BC046307.1 | - | 2, 3 | 364 | 6306542 | ||
BY730445.1 | - | 2, 3 | 415 |