Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019553.2 | NP_062426.2 | 5, 6 | 1203 | ||
GenBank mRNA | AF159131.3 | - | 5, 6 | 1164 | ||
AK010281.1 | - | 5, 6 | 1083 | |||
AK145578.1 | - | 5, 6 | 1187 | |||
AK160095.1 | - | 5, 6 | 1177 | |||
BC030895.1 | - | 4, 5 | 635 | |||
BC043655.1 | - | 5, 6 | 1175 | 5363948 | ||
BC059237.1 | - | 5, 6 | 1163 | 6412782 | ||
BC060220.1 | - | 5, 6 | 1155 |