Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019566.3 | NP_062512.1 | 2 | 837 | ||
GenBank mRNA | AK143623.1 | - | 2 | 830 | ||
AK149848.1 | - | 2 | 830 | |||
AK169853.1 | - | 2 | 821 | |||
AK188210.1 | - | 1 | 315 | |||
BC059775.1 | - | 2 | 816 | 5370916 |