Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cited4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019563.2 | NP_062509.1 | 1 | 983 | ||
GenBank mRNA | AK012136.1 | - | 1 | 975 | ||
AK154030.1 | - | 1 | 974 | |||
BC025116.1 | - | 2 | 655 | |||
BC080723.1 | - | 2 | 740 |