Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019869.3 | NP_063922.2 | 3 | 2073 | ||
GenBank mRNA | AK004928.1 | - | 3 | 2043 | ||
AK050945.1 | - | 3 | 2075 | |||
AK088201.1 | - | 3 | 2039 | |||
AK153230.1 | - | 4 | 1949 | |||
AK167503.1 | - | 3 | 2052 | |||
BC010294.1 | - | 2 | 2768 | 3153900 |