Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Fkbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019873.2 | NP_063926.1 | 2 | 1038 | ||
GenBank mRNA | AK139541.1 | - | 1 | 619 | ||
AK146018.1 | - | 2 | 1108 | |||
BC046338.1 | - | 2 | 1038 | 6395929 |