Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019648.3 | NP_062622.1 | 7 | 1311 | ||
GenBank mRNA | AB003144.1 | - | 6 | 622 | ||
AF434712.1 | - | 6 | 701 | |||
AK076804.1 | - | 6 | 654 | |||
AK090323.1 | - | 6 | 642 | |||
AK147097.1 | - | 6 | 746 | |||
AK159650.1 | - | 6 | 725 | |||
AK163318.1 | - | 6 | 728 | |||
AK165301.1 | - | 7 | 1313 | |||
AK165845.1 | - | 6 | 637 | |||
BC002213.1 | - | 6 | 723 | 3490341 | ||
BC010263.1 | - | 6 | 695 | |||
BC092052.1 | - | 6 | 688 | 3670685 |