Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019648.3 | NP_062622.1 | 3 | 937 | ||
GenBank mRNA | AB003144.1 | - | 2 | 248 | ||
AF434712.1 | - | 2 | 327 | |||
AK076804.1 | - | 2 | 280 | |||
AK090323.1 | - | 2 | 268 | |||
AK147097.1 | - | 2 | 342 | |||
AK159650.1 | - | 2 | 352 | |||
AK163318.1 | - | 2 | 354 | |||
AK165301.1 | - | 3 | 939 | |||
AK165845.1 | - | 2 | 332 | |||
AK189664.1 | - | 1 | 66 | |||
BC002213.1 | - | 2 | 349 | 3490341 | ||
BC010263.1 | - | 2 | 322 | |||
BC092052.1 | - | 2 | 314 | 3670685 |