Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291161.1 | NP_001278090.1 | 2 | 279 | ||
NM_019818.5 | NP_062792.2 | 2 | 360 | |||
GenBank mRNA | AF223950.1 | - | 2 | 326 | ||
AK011745.1 | - | 2 | 312 | |||
AK012130.2 | - | 2 | 314 | |||
AK017389.1 | - | 2 | 313 | |||
AK028132.1 | - | 2 | 312 | |||
AK153837.1 | - | 2 | 315 | |||
BC022610.1 | - | 2 | 296 | |||
BC055279.1 | - | 2 | 324 | 5688754 | ||
BF470824.1 | - | 2 | 336 | |||
CA751902.1 | - | 2 | 252 | |||
Z49228.1 | - | 2 | 86 |