Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019835.2 | NP_062809.2 | 1, 2 | 261 | ||
GenBank mRNA | AB004786.1 | - | 1, 2 | 262 | ||
AF142673.1 | - | 1, 2 | 115 | |||
BE981379.1 | - | 1, 2 | 131 | |||
CJ134591.1 | - | 1, 2 | 262 |