Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019775.3 | NP_062749.2 | 9 | 1028 | ||
GenBank mRNA | AB021968.1 | - | 9 | 976 | ||
AF164524.1 | - | 9 | 1043 | |||
AF186188.1 | - | 9 | 979 | |||
AK004045.1 | - | 9 | 1001 | |||
BC089577.1 | - | 9 | 1000 | 6766672 |