Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025108.1 | NP_001020279.1 | 19 | 3301 | ||
NM_002285.2 | NP_002276.2 | 19 | 3318 | |||
XM_005263943.3 | XP_005264000.2 | 18 | 3195 | |||
XM_005263945.3 | XP_005264002.1 | 10 | 2552 | |||
XM_011511169.2 | XP_011509471.1 | 19 | 3298 | |||
XM_011511170.2 | XP_011509472.1 | 20 | 3437 | |||
XM_011511171.2 | XP_011509473.1 | 19 | 3301 | |||
XM_011511172.2 | XP_011509474.1 | 18 | 3253 | |||
XM_011511173.2 | XP_011509475.1 | 19 | 3314 | |||
XM_011511174.2 | XP_011509476.1 | 19 | 3258 | |||
XM_011511175.2 | XP_011509477.1 | 21 | 3548 | |||
XM_011511176.2 | XP_011509478.1 | 18 | 3239 | |||
XM_011511177.2 | XP_011509479.1 | 19 | 3329 | |||
XM_011511178.2 | XP_011509480.1 | 9 | 2060 | |||
XM_011511179.2 | XP_011509481.1 | 9 | 2057 | |||
XM_017004085.1 | XP_016859574.1 | 10 | 2549 | |||
XM_017004086.1 | XP_016859575.1 | 10 | 3030 | |||
XM_017004087.1 | XP_016859576.1 | 10 | 3027 | |||
GenBank mRNA | AB209634.1 | - | 6 | 1020 | ||
BC136579.1 | - | 17 | 3101 | 9020569 | ||
BC144266.1 | - | 18 | 3176 | 9052786 | ||
CN409428.1 | - | 1 | 94 |