Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sssca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020491.5 | NP_065237.3 | 2 | 93 | ||
GenBank mRNA | AB012865.1 | - | 2 | 61 | ||
AK005265.1 | - | 2 | 65 | |||
AK011957.1 | - | 2 | 83 | |||
AK012592.1 | - | 2 | 71 | |||
AK012704.1 | - | 2 | 71 | |||
BC021593.1 | - | 2 | 63 | 5102521 | ||
BY081504.1 | - | 2 | 151 |