Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028920.1 | - | 5 | 651 | ||
AK004801.1 | - | 5 | 657 | |||
AK030964.1 | - | 1 | 1690 | |||
AK089092.1 | - | 5 | 664 | |||
AK143300.1 | - | 2 | 2441 | |||
BC042602.1 | - | 5 | 645 | |||
BC054433.1 | - | 3 | 343 | |||
BC060170.1 | - | 5 | 642 | |||
BC060209.1 | - | 4 | 1147 | |||
BC078448.1 | - | 5 | 635 | |||
BC125521.1 | - | 5 | 647 | 40130036 | ||
BC137746.1 | - | 5 | 647 | 8860765 |