Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019822.3 | NP_062796.2 | 3 | 352 | ||
GenBank mRNA | AF225959.1 | - | 3 | 268 | ||
AK010919.1 | - | 3 | 329 | |||
AK082084.1 | - | 3 | 335 | |||
AK143843.1 | - | 3 | 327 | |||
AK145474.1 | - | 3 | 314 | |||
AK145792.1 | - | 3 | 343 | |||
AK150091.1 | - | 3 | 305 | |||
AK166878.1 | - | 3 | 341 | |||
AK167085.1 | - | 1 | 740 | |||
BC008974.1 | - | 3 | 352 | 3586698 | ||
BC019746.1 | - | 3 | 281 | 4188750 | ||
BC031517.1 | - | 3 | 295 | 4008987 |