Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019938.3 | NP_064322.1 | 4 | 526 | ||
GenBank mRNA | AB029919.1 | - | 4 | 526 | ||
AK133053.1 | - | 3 | 582 | |||
BC138246.1 | - | 4 | 478 | 8861265 |